Indol-3-acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid

A10556-06 A10556-14 A10556-22 A10556-36
CAAAAA10556-06EA 52 CAD
CAAAAA10556-06 CAAAA10556-14 CAAAA10556-22 CAAAA10556-36
Indol-3-acetic acid 98+%
Indol-3-acetic acid

Plant growth hormone


Purity: 98+% CAS No.: 87-51-4 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Storage Temperature: 2°C to 8°C Flash Point: 171°(339°F) Melting Point: ca 167° dec. Beilstein Reference: 143358 Merck Reference: 14,4964 MDL No.: MFCD00005636

Formula: C₁₀H₉NO₂
Boiling Pt: 120 - 125 °C (0.05 -rr)
Melting Pt: ∼ 167 °C (dec.)
Flash Pt: 171 °C (339 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2
Merck Index: 14,4964

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR