2,6-Dichlorobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10542-14
A10542-22
CAAAAA10542-06EA
59.31
CAD
CAAAAA10542-06
CAAAA10542-14
CAAAA10542-22
2,6-Dichlorobenzoic acid 98%
2,6-Dichlorobenzoic acid
MDL: MFCD00002418
Beilstein Registry No.: 973858
Purity: 98%
CAS No.: 50-30-6
Molecular Formula: C7H4Cl2O2
Formula Weight: 191.01
Melting Point: 141-145°
Beilstein Reference: 973858
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol Boiling Pt: 299 °C (1013 hPa) Melting Pt: 141…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002418 CAS Number: 50-30-6 EINECS: 200-025-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...