Valerophenone 99%
Supplier: Thermo Scientific Chemicals
A10525-14
A10525-22
A10525-36
CAAAA10525-14EA
75.05
CAD
CAAAA10525-14
CAAAA10525-22
CAAAA10525-36
Valerophenone 99%
Valerophenone
MDL: MFCD00009480
Beilstein Registry No.: 1907717
Purity: 99%
CAS No.: 1009-14-9
Molecular Formula: C11H14O
Formula Weight: 162.23
Flash Point: 102°(215°F)
Refractive Index: 1.5143
Melting Point: -9°
Boiling Point: 244-245°
Beilstein Reference: 1907717
MDL No.: MFCD00009480
Formula:
C₆H₅C(O)(CH₂)₃CH₃ Boiling Pt: 244…245 °C Melting Pt: –9 °C Density: 0.978 Flash Pt: 102 °C(215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009480 CAS Number: 1009-14-9 EINECS: 213-767-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...