2,3-Dichlorobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10522-09
A10522-18
CAAAA10522-09EA
109.34
CAD
CAAAA10522-09
CAAAA10522-18
2,3-Dichlorobenzoic acid 98%
2,3-Dichlorobenzoic acid
MDL: MFCD00002413
Beilstein Registry No.: 1946217
Purity: 98%
CAS No.: 50-45-3
Molecular Formula: C7H4Cl2O2
Formula Weight: 191.01
Melting Point: 166-170°
Beilstein Reference: 1946217
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol Melting Pt: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002413 CAS Number: 50-45-3 EINECS: 200-039-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...