Pentaerythritol 98+%

Supplier: Thermo Scientific Chemicals

A10520-0B A10520-0I A10520-30
CAAAA10520-0BEA 68.73 CAD
CAAAA10520-0B CAAAA10520-0I CAAAA10520-30
Pentaerythritol 98+%
Pentaerythritol

MDL: MFCD00004692 Beilstein Registry No.: 1679274


Purity: 98+% CAS No.: 115-77-5 Molecular Formula: C5H12O4 Formula Weight: 136.15 Flash Point: 240°(464°F) Melting Point: 260-264° Boiling Point: 276°/30mm Beilstein Reference: 1679274 Merck Reference: 14,7111 MDL No.: MFCD00004692

Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 276 °C (30 mmHg)
Melting Pt: 260…264 °C
Density: 1.396
Flash Pt: 240 °C (464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 14,7111

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR