2,4-Dichloroaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

A10518-30 A10518-0E A10518-22 A10518-36
CAAAAA10518-30EA 134.3 CAD
CAAAAA10518-30 CAAAA10518-0E CAAAA10518-22 CAAAA10518-36
2,4-Dichloroaniline 99%
2,4-Dichloroaniline

MDL: MFCD00007661 Beilstein Registry No.: 386422


Purity: 99% CAS No.: 554-00-7 Molecular Formula: C6H5Cl2N Formula Weight: 162.02 Flash Point: 149°(300°F) Melting Point: 59-63° Boiling Point: 242° Beilstein Reference: 386422 UN No.: 3442 MDL No.: MFCD00007661

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 242 °C
Melting Pt: 59…63 °C
Density: 1.567
Flash Pt: 149 °C (300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS Number: 554-00-7
EINECS: 209-057-8
UN: 3442
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR