1,4-Dibromobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A10517-18 A10517-30 A10517-0E A10517-22
CAAAAA10517-18EA 43.45 CAD
CAAAAA10517-18 CAAAAA10517-30 CAAAA10517-0E CAAAA10517-22
1,4-Dibromobenzene 98%
1,4-Dibromobenzene

MDL: MFCD00000089 Beilstein Registry No.: 1904543


Purity: 98% CAS No.: 106-37-6 Molecular Formula: C6H4Br2 Formula Weight: 235.91 Flash Point: 100°(212°F) Physical Form: Crystalline powder Density: 1.841 Refractive Index: 1.5742 Appearance: White Odor: Characteristic Melting Point: 86-90° Boiling Point: 219-220° Beilstein Reference: 1904543 Merck Reference: 14,3019 UN No.: 3077 MDL No.: MFCD00000089

Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 219…220 °C
Melting Pt: 86…90 °C
Density: 1.841
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000089
CAS Number: 106-37-6
EINECS: 203-390-2
UN: 3077
ADR: 9,III
Merck Index: 14,3019

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR