3,4-Dibromothiophene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10512-06
A10512-14
CAAAA10512-06EA
121.34
CAD
CAAAA10512-06
CAAAA10512-14
3,4-Dibromothiophene 98+%
3,4-Dibromothiophene
MDL: MFCD00005465
Beilstein Registry No.: 107642
Purity: 98+%
CAS No.: 3141-26-2
Molecular Formula: C4H2Br2S
Formula Weight: 241.94
Flash Point:
Formula:
C₄H₂Br₂S Boiling Pt: 221…222 °C Melting Pt: 4…5 °C Density: 2.188 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005465 CAS Number: 3141-26-2 EINECS: 221-546-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...