4-(Trifluoromethyl)benzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10508-06
A10508-14
A10508-22
CAAAA10508-06EA
39.03
CAD
CAAAA10508-06
CAAAA10508-14
CAAAA10508-22
4-(Trifluoromethyl)benzyl alcohol 98%
4-(Trifluoromethyl)benzyl alcohol
MDL: MFCD00004661
Beilstein Registry No.: 1450174
Purity: 98%
CAS No.: 349-95-1
Molecular Formula: C8H7F3O
Formula Weight: 176.14
Flash Point: 100°(212°F)
Refractive Index: 1.4590
Melting Point: 18-20°
Boiling Point: 78-80°/4mm
Beilstein Reference: 1450174
MDL No.: MFCD00004661
Formula:
C₈H₇F₃O Boiling Pt: 78…80 °C/4 mm Melting Pt: 18…20 °C Density: 1,288 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004661 CAS Number: 349-95-1 EINECS: 206-494-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...