1,3-Dithiane 98%
Supplier: Thermo Scientific Chemicals
A10505-06
A10505-14
A10505-22
CAAAA10505-06EA
75.21
CAD
CAAAA10505-06
CAAAA10505-14
CAAAA10505-22
1,3-Dithiane 98%
1,3-Dithiane
MDL: MFCD00006654
Beilstein Registry No.: 102534
Fieser: 2,182 3,135 4,216 5,287 6,248 7,142 14,157 16,161
Purity: 98%
CAS No.: 505-23-7
Molecular Formula: C4H8S2
Formula Weight: 120.24
Flash Point: 90°(194°F)
Odor: Stench
Boiling Point: 207-208°
Beilstein Reference: 102534
UN No.: 3335
MDL No.: MFCD00006654
Formula:
C₄H₈S₂ Boiling Pt: 207…208 °C Melting Pt: 52…55 °C Density: 1.36 g/cm³ (25 °C) Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006654 CAS Number: 505-23-7 EINECS: 208-006-7 UN: 3335 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...