Benzamide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10501-0E
A10501-22
A10501-36
CAAAA10501-0EEA
592.5
CAD
CAAAA10501-0E
CAAAA10501-22
CAAAA10501-36
Benzamide 98+%
Benzamide
MDL: MFCD00007968
Beilstein Registry No.: 385876
Purity: 98+%
CAS No.: 55-21-0
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 180°(356°F)
Melting Point: 125-129°
Boiling Point: 288°
Beilstein Reference: 385876
Merck Reference: 14,1060
MDL No.: MFCD00007968
Formula:
C₇H₇NO Boiling Pt: 288 °C Melting Pt: 125…129 °C Density: 1.341 Flash Pt: 180 °C(356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007968 CAS Number: 55-21-0 EINECS: 200-227-7 Merck Index: 14,1060 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...