Glutaraldehyde 50% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

A10500-0E A10500-22 A10500-36
CAAAA10500-0EEA 347.6 CAD
CAAAA10500-0E CAAAA10500-22 CAAAA10500-36
Glutaraldehyde 50% in aqueous solution
Glutaraldehyde

MDL: MFCD00007025 Beilstein Registry No.: 605390 Fieser: 1,411


CAS No.: 111-30-8 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.123 Refractive Index: 1.4210 Appearance: Colorless to pale yellow Odor: Stench Melting Point: ca -21° Boiling Point: 100° Beilstein Reference: 605390 Merck Reference: 14,4472 UN No.: 2922 MDL No.: MFCD00007025

Formula: C₅H₈O₂
Boiling Pt: 100 °C
Melting Pt: ∼ 21 °C
Density: 1.123
Storage Temperature: Ambient
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2922
ADR: 8,III
Merck Index: 14,4472

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR