1,2,4-Trimethoxybenzene 98%
Supplier: Thermo Scientific Chemicals
A10483-09
A10483-18
CAAAA10483-09EA
72.05
CAD
CAAAA10483-09
CAAAA10483-18
1,2,4-Trimethoxybenzene 98%
1,2,4-Trimethoxybenzene
MDL: MFCD00008360
Beilstein Registry No.: 2047579
Purity: 98%
CAS No.: 135-77-3
Molecular Formula: C9H12O3
Formula Weight: 168.19
Flash Point:
Formula:
C₉H₁₂O₃ Boiling Pt: 247 °C Melting Pt: 19…21 °C Density: 1.128 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008360 CAS Number: 135-77-3 EINECS: 205-219-7 Merck Index: 14,1073 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...