L(-)-Menthol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol

A10474-0B A10474-18 A10474-30
CAAAA10474-0BEA 472.42 CAD
CAAAA10474-0B CAAAA10474-18 CAAAA10474-30
L(-)-Menthol 99%
L(-)-Menthol

MDL: MFCD00062979 Beilstein Registry No.: 1902293 Fieser: 12,294 13,172 16,203 Optical Rotation: -50°C (c=10 in ethanol)


Purity: 99% CAS No.: 2216-51-5 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 93°(199°F) Melting Point: 42-45° Boiling Point: 212-216° Beilstein Reference: 1902293 Merck Reference: 14,5837 Optical Rotation: -50° (c=10 in ethanol) MDL No.: MFCD00062979

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 212…216 °C
Melting Pt: 42…45 °C
Density: 0.89
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS Number: 2216-51-5
EINECS: 218-690-9
Merck Index: 14,5837

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR