2-Nitro-1,3,5-trimethoxybenzene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,4,6-Trimethoxynitrobenzene, 1,3,5-Trimethoxy-2-nitrobenzene, 1-Nitro-2,4,6-trimethoxybenzene

A10473-22 A10473-06 A10473-14
CAAAAA10473-22EA 2185.14 CAD
CAAAAA10473-22 CAAAA10473-06 CAAAA10473-14
2-Nitro-1,3,5-trimethoxybenzene 98%
2-Nitro-1,3,5-trimethoxybenzene

MDL: MFCD00016992 Beilstein Registry No.: 2277262


Purity: 98% CAS No.: 14227-18-0 Molecular Formula: C9H11NO5 Formula Weight: 213.19 Melting Point: 150-153° Beilstein Reference: 2277262

Formula: C₉H₁₁NO₅
MW: 213.19 g/mol
Melting Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00016992
CAS Number: 14227-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR