1,4-Dibromo-2-nitrobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,5-Dibromonitrobenzene
A10472-14
A10472-22
CAAAA10472-14EA
93.22
CAD
CAAAA10472-14
CAAAA10472-22
1,4-Dibromo-2-nitrobenzene 99%
1,4-Dibromo-2-nitrobenzene
MDL: MFCD00007046
Beilstein Registry No.: 1950425
Purity: 99%
CAS No.: 3460-18-2
Molecular Formula: C6H3Br2NO2
Formula Weight: 280.91
Melting Point: 82-85°
Beilstein Reference: 1950425
Formula:
C₆H₃Br₂NO₂ Melting Pt: 82…85 °C Density: 2.374 Storage Temperature: Ambient |
MDL Number:
MFCD00007046 CAS Number: 3460-18-2 EINECS: 222-404-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...