Sodium gluconate 97%
Supplier: Thermo Scientific Chemicals
A10464-0C
A10464-0E
CAAAA10464-0CEA
565.64
CAD
CAAAA10464-0C
CAAAA10464-0E
Sodium gluconate 97%
Sodium gluconate
MDL: MFCD00064210
Beilstein Registry No.: 3919651
Optical Rotation: +12°C (c=20 in water)
Purity: 97%
CAS No.: 527-07-1
Molecular Formula: C6H11NaO7
Formula Weight: 218.14
Physical Form: Cryst powder
Appearance: White
Odor: Pleasant
Melting Point: ca 206° dec.
Beilstein Reference: 3919651
Merck Reference: 14,4456
Optical Rotation: +12° (c=20 in water)
MDL No.: MFCD00064210
Formula:
HOH₂C(CH(OH))₄COONa Melting Pt: ∼ 206 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064210 CAS Number: 527-07-1 EINECS: 208-407-7 Merck Index: 14,4456 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...