α,α,α-Trifluoro-3-tolualdehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10463-06
A10463-14
A10463-22
CAAAA10463-06EA
84.85
CAD
CAAAA10463-06
CAAAA10463-14
CAAAA10463-22
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde
MDL: MFCD00003373
Beilstein Registry No.: 2327537
Purity: 97%
CAS No.: 454-89-7
Molecular Formula: C8H5F3O
Formula Weight: 174.12
Flash Point: 68°(154°F)
Refractive Index: 1.4650
Odor: Almond-like
Boiling Point: 83-86°/30mm
Beilstein Reference: 2327537
MDL No.: MFCD00003373
Formula:
C₈H₅F₃O Boiling Pt: 83…86 °C/30 mm Density: 1.304 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003373 CAS Number: 454-89-7 EINECS: 207-228-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...