2,6-Difluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A10454-18 A10454-30
CAAAA10454-18EA 154.52 CAD
CAAAA10454-18 CAAAA10454-30
2,6-Difluorobenzonitrile 98%
2,6-Difluorobenzonitrile

MDL: MFCD00001778 Beilstein Registry No.: 2045292


Purity: 98% CAS No.: 1897-52-5 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 80°(176°F) Refractive Index: 1.4875 Odor: Almond-like Melting Point: 28-32° Boiling Point: 197-198° Beilstein Reference: 2045292 UN No.: 3439 MDL No.: MFCD00001778

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 197…198 °C
Melting Pt: 28…32 °C
Density: 1.246
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001778
CAS Number: 1897-52-5
EINECS: 217-589-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR