Ethyl bromoacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10448-0E
A10448-22
A10448-36
CAAAA10448-0EEA
663.6
CAD
CAAAA10448-0E
CAAAA10448-22
CAAAA10448-36
Ethyl bromoacetate 98%
Ethyl bromoacetate
MDL: MFCD00000191
Purity: 98%
CAS No.: 105-36-2
Molecular Formula: C4H7BrO2
Formula Weight: 167.00
Flash Point: 47°(117°F)
Physical Form: Clear liquid
Density: 1.506
Refractive Index: 1.4510
Appearance: Colorless
Melting Point: -38°
Boiling Point: 159°
UN No.: 1603
MDL No.: MFCD00000191
Formula:
BrCH₂COOCH₂CH₃ Boiling Pt: 159 °C Melting Pt: –38 °C Density: 1.506 Flash Pt: 47 °C (117 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000191 CAS Number: 105-36-2 EINECS: 203-290-9 UN: 1603 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...