3,5-Dihydroxytoluene monohydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

A10440-06 A10440-14 A10440-22
CAAAA10440-06EA 50.56 CAD
CAAAA10440-06 CAAAA10440-14 CAAAA10440-22
3,5-Dihydroxytoluene monohydrate 97%
3,5-Dihydroxytoluene monohydrate

MDL: MFCD00149092 Beilstein Registry No.: 5103474


Purity: 97% CAS No.: 6153-39-5 Molecular Formula: C7H8O2.H2O Formula Weight: 142.16 (124.14anhy) Melting Point: 56-60° Boiling Point: 147°/5mm Merck Reference: 14,6864 MDL No.: MFCD00149092

Formula: C₇H₈O₂·1H₂O
Boiling Pt: 147 °C/5 mm
Melting Pt: 56…60 °C
Density: 1.29 g/cm³ (4 °C)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00149092
CAS Number: 6153-39-5
EINECS: 207-984-2
Merck Index: 14,6864

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR