4'-Hydroxy-3'-methoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin

A10439-36 A10439-18 A10439-30
CAAAAA10439-36EA 341.28 CAD
CAAAAA10439-36 CAAAA10439-18 CAAAA10439-30
4'-Hydroxy-3'-methoxyacetophenone 98%
4'-Hydroxy-3'-methoxyacetophenone

MDL: MFCD00008747 Beilstein Registry No.: 637373


Purity: 98% CAS No.: 498-02-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 112-116° Boiling Point: 295-300° Merck Reference: 14,741 MDL No.: MFCD00008747

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 295…300 °C
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00008747
CAS Number: 498-02-2
EINECS: 207-854-5
Merck Index: 14,741

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR