1-Methyl-2-thiourea 97+%, REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10437-14
A10437-22
CAAAA10437-14EA
84.06
CAD
CAAAA10437-14
CAAAA10437-22
1-Methyl-2-thiourea 97+%, REacton®
1-Methyl-2-thiourea
MDL: MFCD00004938
Beilstein Registry No.: 506162
Purity: 98+%
CAS No.: 598-52-7
Molecular Formula: C2H6N2S
Formula Weight: 90.15
pH: 7
Melting Point: 119-124°
Beilstein Reference: 506162
UN No.: 2811
MDL No.: MFCD00004938
Formula:
C₂H₆N₂S Melting Pt: 119…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004938 CAS Number: 598-52-7 EINECS: 209-936-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...