L(+)-Cysteine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
MDL: MFCD00064306
Beilstein Registry No.: 1721408
Optical Rotation: +7.6°C (c=5 in 5N HCl)
Purity: 98+%
CAS No.: 52-90-4
Molecular Formula: C3H7NO2S
Formula Weight: 121.16
Physical Form: Crystalline powder
Sensitivity: Air Sensitive
Appearance: White
Odor: Odorless
Melting Point: ca 220° dec.
Beilstein Reference: 1721408
Merck Reference: 14,2781
Optical Rotation: +7.9° (c=5 in 5N HCl)
MDL No.: MFCD00064306
Formula:
HSCH₂CH(NH₂)CO₂H MW: 121.16 g/mol Boiling Pt: 254 Melting Pt: 115 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064306 CAS Number: 52-90-4 EINECS: 200-158-2 Merck Index: 14,2781 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...