9-Bromofluorene 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10428-06 A10428-18
CAAAAA10428-06EA 59.88 CAD
CAAAAA10428-06 CAAAA10428-18 CAAAA10428-30
9-Bromofluorene 98+%
9-Bromofluorene

MDL: MFCD00001133 Beilstein Registry No.: 2047220


Purity: 98+% CAS No.: 1940-57-4 Molecular Formula: C13H9Br Formula Weight: 245.12 Odor: Irritating Melting Point: 101-105° Beilstein Reference: 2047220 UN No.: 3261 MDL No.: MFCD00001133

Formula: C₁₃H₉Br
Boiling Pt: 318 °C (1013 hPa)
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00001133
CAS Number: 1940-57-4
EINECS: 217-722-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR