5-Nitroanthranilic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-5-nitrobenzoic acid
A10426-09
A10426-18
CAAAA10426-09EA
89.74
CAD
CAAAA10426-09
CAAAA10426-18
CAAAA10426-30
5-Nitroanthranilic acid 98%
5-Nitroanthranilic acid
MDL: MFCD00017039
Beilstein Registry No.: 646219
Purity: 98%
CAS No.: 616-79-5
Molecular Formula: C7H6N2O4
Formula Weight: 182.14
Melting Point: ca 270° dec.
Beilstein Reference: 646219
Formula:
C₇H₆N₂O₄ MW: 182.14 g/mol Melting Pt: ∼ 270 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00017039 CAS Number: 616-79-5 EINECS: 210-493-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...