Azobenzene 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10425-14
A10425-22
A10425-36
CAAAA10425-14EA
97.49
CAD
CAAAA10425-14
CAAAA10425-22
CAAAA10425-36
Azobenzene 97+%
Azobenzene
MDL: MFCD00003022
Purity: 97+%
CAS No.: 103-33-3
Molecular Formula: C12H10N2
Formula Weight: 182.23
Melting Point: 65-69°
Boiling Point: 293°
Merck Reference: 14,917
UN No.: 3077
MDL No.: MFCD00003022
Formula:
C₁₂H₁₀N₂ Boiling Pt: 293 °C Melting Pt: 65…69 °C Density: 1.203 Flash Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003022 CAS Number: 103-33-3 EINECS: 203-102-5 UN: 3077 ADR: 9,III Merck Index: 14,917 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...