Pyridine-N-oxide 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Pyridine 1-oxide, Pyridine N-oxide
A10419-36
A10419-14
A10419-22
CAAAAA10419-36EA
418.7
CAD
CAAAAA10419-36
CAAAA10419-14
CAAAA10419-22
Pyridine-N-oxide 95%
Pyridine N-oxide
MDL: MFCD00006194
Beilstein Registry No.: 105257
Fieser: 1,966 2,353 5,567 9,396
Purity: 95%
CAS No.: 694-59-7
Molecular Formula: C5H5NO
Formula Weight: 95.10
Flash Point: 143°(289°F)
Melting Point: 61-66°
Boiling Point: 270°
Beilstein Reference: 105257
Merck Reference: 14,7972
MDL No.: MFCD00006194
Formula:
C₅H₅NO MW: 95.1 g/mol Boiling Pt: 270 Melting Pt: 61…66 °C Flash Pt: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006194 CAS Number: 694-59-7 EINECS: 211-774-6 Merck Index: 14,7972 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...