2,2,3,3,4,4,4-Heptafluoro-1-butanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol

A10414-14 A10414-30 A10414-09 A10414-18
CAAAAA10414-14EA 237 CAD
CAAAAA10414-14 CAAAAA10414-30 CAAAA10414-09 CAAAA10414-18
2,2,3,3,4,4,4-Heptafluoro-1-butanol 98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol

MDL: MFCD00004674 Beilstein Registry No.: 1761907


Purity: 98% CAS No.: 375-01-9 Molecular Formula: C4H3F7O Formula Weight: 200.06 Flash Point: 25°(77°F) Refractive Index: 1.300 Boiling Point: 95-97° Beilstein Reference: 1761907 UN No.: 1987 MDL No.: MFCD00004674

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 95…97 °C
Density: 1.596
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004674
CAS Number: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR