L(+)-Histidine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

A10413-14 A10413-22 A10413-36
CAAA-A10413-14EA 57.35 CAD
CAAA-A10413-14 CAAA-A10413-22 CAAA-A10413-36
L(+)-Histidine 98+%
L(+)-Histidine

MDL: MFCD00064315 Beilstein Registry No.: 4673585 Optical Rotation: -39°C (c=5 in water)

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: ∼ 281 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3
Merck Index: 14,4720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR