Ninhydrin 99%

Supplier: Thermo Scientific Chemicals
Warning

A10409-14 A10409-09 A10409-18 A10409-30
CAAAAA10409-14EA 99.7 CAD
CAAAAA10409-14 CAAAA10409-09 CAAAA10409-18 CAAAA10409-30
Ninhydrin 99%
Ninhydrin

MDL: MFCD00003791 Beilstein Registry No.: 1910963 Fieser: 1,732 4,356

Formula: C₉H₆O₄
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 251 °C (dec.)
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 14,6554
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR