Methyl-2,5-dihydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Gentisic acid methyl ester, Methyl 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid methyl ester

A10407-06 A10407-14
CAAAA10407-03EA 73.45 CAD
CAAAA10407-03 CAAAA10407-06 CAAAA10407-14
Methyl-2,5-dihydroxybenzoate 98%
Methyl 2,5-dihydroxybenzoate

MDL: MFCD00016464


Purity: 98% CAS No.: 2150-46-1 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 86-87° MDL No.: MFCD00016464

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 86…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00016464
CAS Number: 2150-46-1
EINECS: 218-427-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR