4-Pentyn-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10405-06
A10405-14
A10405-22
CAAAA10405-06EA
83.58
CAD
CAAAA10405-06
CAAAA10405-14
CAAAA10405-22
4-Pentyn-1-ol 97%
4-Pentyn-1-ol
Purity: 97%
CAS No.: 5390-04-5
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 62°(143°F)
Refractive Index: 1.4450
Boiling Point: 153-155°
Beilstein Reference: 1736712
MDL No.: MFCD00002974
Formula:
C₅H₈O Boiling Pt: 153…155 °C Density: 0.907 Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002974 CAS Number: 5390-04-5 EINECS: 226-383-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...