Dimethyl fumarate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fumaric acid dimethyl ester

A10402-14 A10402-22 A10402-30
CAAAAA10402-14EA 47.4 CAD
CAAAAA10402-14 CAAAAA10402-22 CAAAA10402-30
Dimethyl fumarate 99%
Dimethyl fumarate

MDL: MFCD00064438 Beilstein Registry No.: 774590


Purity: 99% CAS No.: 624-49-7 Molecular Formula: C6H8O4 Formula Weight: 144.13 Melting Point: 102-105° Boiling Point: 192-193° Merck Reference: 14,4287 MDL No.: MFCD00064438

Formula: C₆H₈O₄
Boiling Pt: 192…193 °C
Melting Pt: 102…105 °C
Density: 1.37
Storage Temperature: Ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 14,4287

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR