2-Nitrobenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

A10393-0E A10393-22 A10393-36
CAAAA10393-0EEA 687.3 CAD
CAAAA10393-0E CAAAA10393-22 CAAAA10393-36
2-Nitrobenzoic acid 95%
2-Nitrobenzoic acid

MDL: MFCD00007137 Beilstein Registry No.: 1910632


Purity: 95% CAS No.: 552-16-9 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 146-149° Beilstein Reference: 1910632 Merck Reference: 14,6589 MDL No.: MFCD00007137

Formula: C₇H₅NO₄
Melting Pt: 146…149 °C
Density: 1.58
Storage Temperature: Ambient
MDL Number: MFCD00007137
CAS Number: 552-16-9
EINECS: 209-004-9
Merck Index: 14,6589

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR