1,12-Dodecanedioic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecanedioic acid, 1,10-Decanedicarboxylic acid
A10387-0B
A10387-30
CAAAA10387-0BEA
346.02
CAD
CAAAA10387-0B
CAAAA10387-30
1,12-Dodecanedioic acid 99%
1,12-Dodecanedioic acid
MDL: MFCD00002735
Beilstein Registry No.: 1782580
Purity: 99%
CAS No.: 693-23-2
Molecular Formula: HO2C(CH2)10CO2H
Formula Weight: 230.31
Flash Point: 227°(440°F)
Odor: Odorless
Boiling Point: 244-245°/10mm
Beilstein Reference: 1782580
MDL No.: MFCD00002735
Formula:
C₁₂H₂₂O₄ Boiling Pt: 244…245 °C/10 mm Melting Pt: 127…131 °C Density: 1.150 Flash Pt: 227 °C(440 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002735 CAS Number: 693-23-2 EINECS: 211-746-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...