2,3-Dimethylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A10383-06 A10383-14 A10383-22
CAAAA10383-06EA 69.2 CAD
CAAAA10383-06 CAAAA10383-14 CAAAA10383-22
2,3-Dimethylbenzoic acid 98%
2,3-Dimethylbenzoic acid

MDL: MFCD00002479 Beilstein Registry No.: 971590


Purity: 98% CAS No.: 603-79-2 Molecular Formula: C9H10O2 Formula Weight: 150.18 Odor: Odorless Melting Point: 143-147° Beilstein Reference: 971590 MDL No.: MFCD00002479

Formula: C₉H₁₀O₂
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 143…147 °C
Density: 1,283 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002479
CAS Number: 603-79-2
EINECS: 210-058-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR