3-Pyridylmethanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol
A10381-0B
A10381-18
A10381-30
CAAAA10381-0BEA
899.02
CAD
CAAAA10381-0B
CAAAA10381-18
CAAAA10381-30
3-Pyridylmethanol 98%
3-Pyridylmethanol
MDL: MFCD00006407
Beilstein Registry No.: 107851
Purity: 98%
CAS No.: 100-55-0
Molecular Formula: C6H7NO
Formula Weight: 109.13
Storage Temperature: 2°C to 8°C
Flash Point: 149°(300°F)
Refractive Index: 1.5450
Melting Point: -8 to -6°
Boiling Point: 154°/28mm
Beilstein Reference: 107851
Merck Reference: 14,6527
MDL No.: MFCD00006407
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 154 °C/28 mm Melting Pt: –8…–6 °C Density: 1.130 Flash Pt: 149 °C (300 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006407 CAS Number: 100-55-0 EINECS: 202-864-6 Merck Index: 14,6527 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...