2-(Methylthio)benzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10379-06
A10379-14
CAAAA10379-06EA
162.74
CAD
CAAAA10379-06
CAAAA10379-14
2-(Methylthio)benzoic acid 98+%
2-(Methylthio)benzoic acid
MDL: MFCD00029934
Purity: 98+%
CAS No.: 3724-10-5
Molecular Formula: C8H8O2S
Formula Weight: 168.21
Melting Point: 166-168°
MDL No.: MFCD00029934
Formula:
C₈H₈O₂S Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00029934 CAS Number: 3724-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...