1,2-Difluorobenzene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10377-09
A10377-18
A10377-30
CAAAA10377-09EA
87.53
CAD
CAAAA10377-09
CAAAA10377-18
CAAAA10377-30
1,2-Difluorobenzene 98+%
1,2-Difluorobenzene
MDL: MFCD00000284
Beilstein Registry No.: 1905113
Purity: 98+%
CAS No.: 367-11-3
Molecular Formula: C6H4F2
Formula Weight: 114.09
Flash Point: 1°(33°F)
Refractive Index: 1.4440
Odor: Characteristic
Melting Point: -34°
Boiling Point: 92-94°
Beilstein Reference: 1905113
UN No.: 1993
MDL No.: MFCD00000284
Formula:
C₆H₄F₂ Boiling Pt: 92…94 °C Melting Pt: –34 °C Density: 1.171 Flash Pt: 1 °C (33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000284 CAS Number: 367-11-3 EINECS: 206-680-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...