2-Ethylhexylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10372-0F
A10372-AE
A10372-AP
CAAAA10372-0FEA
285.98
CAD
CAAAA10372-0F
CAAAA10372-AE
CAAAA10372-AP
2-Ethylhexylamine 98%
2-Ethylhexylamine
MDL: MFCD00008148
Beilstein Registry No.: 1209249
Purity: 98%
CAS No.: 104-75-6
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 50°(122°F)
Refractive Index: 1.4300
Odor: Amine-like
Melting Point: -76°
Boiling Point: 169°
Beilstein Reference: 1209249
UN No.: 2276
MDL No.: MFCD00008148
Formula:
C₈H₁₉N Boiling Pt: 169 °C Melting Pt: –76 °C Density: 0.789 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008148 CAS Number: 104-75-6 EINECS: 203-233-8 UN: 2276 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...