Trityl alcohol 98%
Supplier: Thermo Scientific Chemicals
A10366-0B
A10366-18
A10366-30
CAAAA10366-0BEA
834.24
CAD
CAAAA10366-0B
CAAAA10366-18
CAAAA10366-30
Trityl alcohol 98%
Trityl alcohol
MDL: MFCD00004445
Beilstein Registry No.: 1460837
Purity: 98%
CAS No.: 76-84-6
Molecular Formula: (C6H5)3COH
Formula Weight: 260.34
Physical Form: Cryst powder
Appearance: White to cream
Melting Point: 161-164°
Boiling Point: 360-380°
Beilstein Reference: 1460837
Merck Reference: 14,9739
MDL No.: MFCD00004445
Formula:
C₁₉H₁₆O Boiling Pt: 360…380 °C Melting Pt: 161…164 °C Density: 1.1884 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004445 CAS Number: 76-84-6 EINECS: 200-988-5 Merck Index: 14,9739 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...