4-Fluorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Fluoro-4-methylbenzene
A10365-22
A10365-36
CAAAA10365-22EA
62.88
CAD
CAAAA10365-22
CAAAA10365-36
4-Fluorotoluene 99%
4-Fluorotoluene
MDL: MFCD00000358
Beilstein Registry No.: 1362373
Purity: 99%
CAS No.: 352-32-9
Molecular Formula: C7H7F
Formula Weight: 110.13
Flash Point: 10°(50°F)
Refractive Index: 1.4685
Odor: Aromatic
Melting Point: -56°
Boiling Point: 115-116°
Beilstein Reference: 1362373
Merck Reference: 14,4180
UN No.: 2388
MDL No.: MFCD00000358
Formula:
C₇H₇F MW: 110.13 g/mol Boiling Pt: 115…116 °C Melting Pt: –56 °C Density: 1.001 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000358 CAS Number: 352-32-9 EINECS: 206-520-6 UN: 2388 ADR: 3,II Merck Index: 14,4180 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...