Morpholine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10355-0E
A10355-22
A10355-36
CAAAA10355-0EEA
199.08
CAD
CAAAA10355-0E
CAAAA10355-22
CAAAA10355-36
Morpholine 99%
Morpholine
MDL: MFCD00005972
Beilstein Registry No.: 102549
Fieser: 1,705 11,352
Purity: 99%
CAS No.: 110-91-8
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 1.007
Refractive Index: 1.4540
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Amine-like
Melting Point: -5°
Boiling Point: 129°
Beilstein Reference: 102549
Merck Reference: 14,6277
UN No.: 2054
MDL No.: MFCD00005972
Formula:
C₄H₉NO Boiling Pt: 129 °C Melting Pt: –5 °C Density: 1.007 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005972 CAS Number: 110-91-8 EINECS: 203-815-1 UN: 2054 ADR: 8,I Merck Index: 14,6277 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...