3',5'-Bis(trifluoromethyl)acetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10341-06
A10341-14
CAAAA10341-03EA
64.18
CAD
CAAAA10341-03
CAAAA10341-06
CAAAA10341-14
3',5'-Bis(trifluoromethyl)acetophenone 98%
3',5'-Bis(trifluoromethyl)acetophenone
MDL: MFCD00009910
Beilstein Registry No.: 2384789
Purity: 98%
CAS No.: 30071-93-3
Molecular Formula: C10H6F6O
Formula Weight: 256.15
Flash Point: 82°(179°F)
Refractive Index: 1.4230
Boiling Point: 85°/8mm
Beilstein Reference: 2384789
MDL No.: MFCD00009910
Formula:
C₁₀H₆F₆O Boiling Pt: 85 °C/8 mm Density: 1.422 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009910 CAS Number: 30071-93-3 EINECS: 250-023-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...