4-Ethoxybenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

A10331-09 A10331-18
CAAAA10331-09EA 156.1 CAD
CAAAA10331-09 CAAAA10331-18
4-Ethoxybenzyl alcohol 98%
4-Ethoxybenzyl alcohol

MDL: MFCD00004655


Purity: 98% CAS No.: 6214-44-4 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 110°(230°F) Refractive Index: 1.5320 Melting Point: 32-34° Boiling Point: 272-274° MDL No.: MFCD00004655

Formula: C₉H₁₂O₂
Boiling Pt: 272…274 °C
Melting Pt: 32…34 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004655
CAS Number: 6214-44-4
EINECS: 228-283-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR