2,4,5-Trimethyloxazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,4,5-Trimethyl-1,3-oxazole
A10328-06
A10328-14
CAAAA10328-06EA
49.93
CAD
CAAAA10328-06
CAAAA10328-14
2,4,5-Trimethyloxazole 97%
2,4,5-Trimethyloxazole
MDL: MFCD00005308
Beilstein Registry No.: 107043
Purity: 97%
CAS No.: 20662-84-4
Molecular Formula: C6H9NO
Formula Weight: 111.14
Storage Temperature: 2°C to 8°C
Flash Point: 33°(91°F)
Refractive Index: 1.4420
Odor: Pungent
Boiling Point: 133-134°
Beilstein Reference: 107043
UN No.: 1993
MDL No.: MFCD00005308
Formula:
C₆H₉NO MW: 111.14 g/mol Boiling Pt: 133…134 °C Density: 0.964 Flash Pt: 33 °C(91 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005308 CAS Number: 20662-84-4 EINECS: 243-952-4 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...