Methyl chloroacetate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10325-30
A10325-0E
CAAAAA10325-30EA
37.76
CAD
CAAAAA10325-30
CAAAA10325-0E
Methyl chloroacetate 99+%
Methyl chloroacetate
MDL: MFCD00000931
Beilstein Registry No.: 506255
Purity: 99+%
CAS No.: 96-34-4
Molecular Formula: ClCH2CO2CH3
Formula Weight: 108.52
Flash Point: 51°(123°F)
Refractive Index: 1.4220
Melting Point: -32°
Boiling Point: 128-130°
Beilstein Reference: 506255
Merck Reference: 14,6042
UN No.: 2295
MDL No.: MFCD00000931
Formula:
C₃H₅ClO₂ Boiling Pt: 128…130 °C Melting Pt: –32 °C Density: 1.236 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000931 CAS Number: 96-34-4 EINECS: 202-501-1 UN: 2295 ADR: 6.1,I Merck Index: 14,6042 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...