4,4'-Isopropylidenediphenol 97+%

Supplier: Thermo Scientific Chemicals
Danger

A10324-0B A10324-0I A10324-30
CAAAA10324-0BEA 90.85 CAD
CAAAA10324-0B CAAAA10324-0I CAAAA10324-30
4,4'-Isopropylidenediphenol 97+%
4,4'-Isopropylidenediphenol

MDL: MFCD00002366 Beilstein Registry No.: 1107700


Purity: 97+% CAS No.: 80-05-7 Molecular Formula: C15H16O2 Formula Weight: 228.29 Flash Point: 227°(440°F) Melting Point: 153-158° Boiling Point: 200°/4mm Beilstein Reference: 1107700 Merck Reference: 14,1297 MDL No.: MFCD00002366

Formula: C₁₅H₁₆O₂
Boiling Pt: 200 °C/4 mm
Melting Pt: 153…158 °C
Density: 1.205 g/cm³ (20 °C)
Flash Pt: 227 °C (440 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002366
CAS Number: 80-05-7
EINECS: 201-245-8
UN: 3077
ADR: 9,III
Merck Index: 14,1297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR