n-Hexadecane 99%

Supplier: Thermo Scientific Chemicals
Danger

A10322-0F A10322-AE A10322-AP
CAAAA10322-0FEA 931.99 CAD
CAAAA10322-0F CAAAA10322-AE CAAAA10322-AP
n-Hexadecane 99%
n-Hexadecane

Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.


Purity: 99% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colourless Melting Point: 18° Boiling Point: 287° Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water. Beilstein Reference: 1736592 MDL No.: MFCD00008998


Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142

Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C
Melting Pt: 18 °C
Density: 0.773
Flash Pt: 135 °C(275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR